siRNA Details
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039025.2 | NP_001034114.2 | 26, 27 | 3924 | ||
XM_008763674.1 | XP_008761896.1 | 27, 28 | 3743 | |||
XM_008763675.1 | XP_008761897.1 | 28, 29 | 5457 | |||
GenBank mRNA | BC100262.1 | - | 7, 8 | 1046 | 7373937 |
