siRNA Details
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177932.2 | NP_808793.1 | 1 | 87 | ||
XM_006239127.2 | XP_006239189.1 | 2 | 388 | |||
XM_008764144.1 | XP_008762366.1 | 2 | 390 | |||
GenBank mRNA | AY225592.1 | - | 1 | 87 |
