siRNA Details
Gene Symbol: Prdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077648.1 | NP_001071116.1 | 8 | 2866 | ||
XM_006239335.2 | XP_006239397.1 | 9 | 2949 | |||
XM_006239337.2 | XP_006239399.1 | 5 | 2536 | |||
XM_008764255.1 | XP_008762477.1 | 8 | 6500 | |||
GenBank mRNA | U17837.1 | - | 8 | 2866 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001256380, NM_001081355, BC151062.
Gene Ontology Categories:
