siRNA Details
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108005.1 | NP_001101475.1 | 2 | 189 | ||
XM_006238671.2 | XP_006238733.1 | 2 | 180 | |||
XM_006238674.2 | XP_006238736.1 | 2 | 180 | |||
XM_006238675.2 | XP_006238737.1 | 2 | 180 |
