siRNA Details
Gene Symbol: RGD1562284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134557.2 | NP_001128029.1 | 2 | 206 | ||
XM_006239632.2 | XP_006239694.1 | 3 | 571 | |||
GenBank mRNA | FQ228724.1 | - | 3 | 362 | ||
FQ229106.1 | - | 3 | 365 |
