siRNA Details
Gene Symbol: Cmpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108017.1 | NP_001101487.1 | 5 | 1323 | ||
XM_006239938.2 | XP_006240000.1 | 6 | 2010 | |||
XM_006239939.2 | XP_006240001.1 | 6 | 2010 |
