siRNA Details
Gene Symbol: Dcaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100718.1 | NP_001094188.1 | 7 | 917 | ||
XM_006240279.2 | XP_006240341.1 | 7 | 719 | |||
XM_008764782.1 | XP_008763004.1 | 7 | 772 | |||
GenBank mRNA | BC079428.1 | - | 2 | 36 |
