siRNA Details
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034929.1 | NP_001030101.1 | 4 | 789 | ||
XM_006240283.2 | XP_006240345.1 | 4 | 1093 | |||
XM_008764784.1 | XP_008763006.1 | 3 | 1222 | |||
GenBank mRNA | BC105849.1 | - | 4 | 789 | 7457541 |
