siRNA Details
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199394.1 | NP_955426.1 | 8 | 836 | ||
XM_006240343.2 | XP_006240405.1 | 9 | 1524 | |||
XM_006240344.1 | XP_006240406.1 | 9 | 884 | |||
XM_006240345.2 | XP_006240407.1 | 9 | 1499 | |||
XM_006240346.2 | XP_006240408.1 | 8 | 983 | |||
GenBank mRNA | BC062044.1 | - | 8 | 836 | 5597848 |
