siRNA Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108052.1 | NP_001101522.1 | 5 | 1573 | ||
XM_006240473.2 | XP_006240535.1 | 5 | 1842 | |||
XM_006240474.2 | XP_006240536.1 | 6 | 1630 | |||
XM_008764804.1 | XP_008763026.1 | 5 | 6220 |
