siRNA Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108052.1 | NP_001101522.1 | 3 | 416 | ||
XM_006240473.2 | XP_006240535.1 | 3 | 685 | |||
XM_006240474.2 | XP_006240536.1 | 4 | 473 | |||
XM_008764804.1 | XP_008763026.1 | 3 | 5063 |
