siRNA Details
Gene Symbol: Bcl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277287.1 | NP_001264216.1 | 4 | 1299 | ||
NM_001277288.1 | NP_001264217.1 | 3 | 1095 | |||
XM_006240540.2 | XP_006240602.1 | 3 | 1258 | |||
GenBank mRNA | JF520428.1 | - | 3 | 1095 | ||
JF520429.1 | - | 4 | 1299 |
