siRNA Details
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014080.1 | NP_001014102.1 | 15, 16 | 1911 | ||
XM_006256612.2 | XP_006256674.1 | 16, 17 | 1896 | |||
XM_006256613.2 | XP_006256675.1 | 15, 16 | 1898 | |||
GenBank mRNA | BC085791.1 | - | 15, 16 | 1911 | 7109118 | |
FQ217276.1 | - | 15, 16 | 1889 |
