siRNA Details
Gene Symbol: Tjp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108073.1 | NP_001101543.1 | 4 | 394 | ||
XM_006240932.2 | XP_006240994.1 | 4 | 277 | |||
XM_006240933.2 | XP_006240995.1 | 4 | 740 |
