siRNA Details
Gene Symbol: Spic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108080.1 | NP_001101550.1 | 5 | 701 | ||
XM_006241223.2 | XP_006241285.1 | 6 | 880 | |||
XM_006241224.2 | XP_006241286.1 | 6 | 857 | |||
XM_006241225.2 | XP_006241287.1 | 5 | 784 | |||
XM_006241226.2 | XP_006241288.1 | 5 | 760 | |||
GenBank mRNA | FQ233971.1 | - | 6 | 811 |
