siRNA Details
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135858.1 | NP_001129330.1 | 15 | 2498 | ||
XM_008765316.1 | XP_008763538.1 | 14 | 2408 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135858.1 | NP_001129330.1 | 15 | 2498 | ||
XM_008765316.1 | XP_008763538.1 | 14 | 2408 |