siRNA Details
Gene Symbol: Pkhd1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034931.2 | NP_001030103.2 | 69 | 11185 | ||
GenBank mRNA | DY546293.1 | - | 1 | 139 | ||
FM120324.1 | - | 2 | 296 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pkhd1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034931.2 | NP_001030103.2 | 69 | 11185 | ||
GenBank mRNA | DY546293.1 | - | 1 | 139 | ||
FM120324.1 | - | 2 | 296 |