siRNA Details
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014085.1 | NP_001014107.1 | 3 | 474 | ||
XM_006241811.1 | XP_006241873.1 | 3 | 549 | |||
XM_006241812.1 | XP_006241874.1 | 3 | 518 | |||
GenBank mRNA | BC083712.1 | - | 3 | 474 | 7190634 |
