siRNA Details
Gene Symbol: LOC100911804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750381.3 | XP_003750429.1 | 1457 | |||
XM_006241953.2 | XP_006242015.1 | 1554 | ||||
XM_006241954.2 | XP_006242016.1 | 1436 | ||||
XM_006241955.2 | XP_006242017.1 | 4 | 1271 | |||
XM_006241956.1 | XP_006242018.1 | 1006 | ||||
XM_006241957.2 | XP_006242019.1 | 4 | 1251 | |||
XM_006241958.2 | XP_006242020.1 | 5 | 1327 | |||
XM_006241959.2 | XP_006242021.1 | 5 | 1299 | |||
XM_008765799.1 | XP_008764021.1 | 1026 | ||||
XM_008765800.1 | XP_008764022.1 | 5 | 1279 | |||
XM_008765801.1 | XP_008764023.1 | 2 | 866 |
Gene Symbol: Apol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241942.2 | XP_006242004.1 | 4 | 1553 | ||
XM_006241943.2 | XP_006242005.1 | 5 | 1435 | |||
XM_006241944.2 | XP_006242006.1 | 4 | 1270 | |||
XM_006241945.1 | XP_006242007.1 | 4 | 1006 | |||
XM_006241946.2 | XP_006242008.1 | 4 | 1250 | |||
XM_006241947.2 | XP_006242009.1 | 5 | 1326 | |||
XM_006241948.2 | XP_006242010.1 | 5 | 1298 | |||
XM_008765644.1 | XP_008763866.1 | 4 | 1026 | |||
XM_008765645.1 | XP_008763867.1 | 5 | 1278 | |||
XM_008765646.1 | XP_008763868.1 | 2 | 866 | |||
XM_235465.8 | XP_235465.6 | 5 | 1456 |
