siRNA Details
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750385.3 | XP_003750433.1 | 11 | 7955 | ||
XM_006242130.2 | XP_006242192.1 | 12 | 3012 | |||
XM_008765806.1 | XP_008764028.1 | 11 | 3625 | |||
XM_235497.8 | XP_235497.6 | 12 | 2686 | |||
GenBank mRNA | AB588919.1 | - | 12 | 2573 | ||
AB588920.1 | - | 11 | 2428 | |||
AB588921.1 | - | 12 | 2555 |
