siRNA Details
Gene Symbol: Prickle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199396.1 | NP_955428.1 | 7 | 1889 | ||
XM_006242213.2 | XP_006242275.1 | 6 | 1322 | |||
XM_006242214.2 | XP_006242276.1 | 7 | 3786 | |||
XM_006242215.2 | XP_006242277.1 | 7 | 1717 | |||
GenBank mRNA | AF399843.1 | - | 4 | 1078 |
