siRNA Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108112.1 | NP_001101582.1 | 9 | 1035 | ||
XM_006257400.2 | XP_006257462.1 | 9 | 982 | |||
GenBank mRNA | BC166473.1 | - | 9 | 935 | 9090730 | |
FQ231766.1 | - | 1 | 72 | |||
FQ234782.1 | - | 1 | 72 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765882.1 | XP_008764104.1 | 7 | 611 |
