siRNA Details
Gene Symbol: Espl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170602.1 | NP_001164073.1 | 26 | 5566 | ||
XM_006242460.2 | XP_006242522.1 | 26 | 5711 | |||
XM_006242461.2 | XP_006242523.1 | 26 | 5706 |
