siRNA Details
Gene Symbol: Espl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170602.1 | NP_001164073.1 | 10 | 2165 | ||
XM_006242460.2 | XP_006242522.1 | 10 | 2310 | |||
XM_006242461.2 | XP_006242523.1 | 10 | 2305 |
Gene Symbol: LOC103690081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765876.1 | XP_008764098.1 | 8 | 1763 |
