siRNA Details
Gene Symbol: RGD1561444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242701.2 | XP_006242763.1 | 4 | 3258 | ||
XM_006242702.2 | XP_006242764.1 | 3 | 3351 | |||
XM_008766018.1 | XP_008764240.1 | 3 | 2952 | |||
XM_235936.8 | XP_235936.7 | 3 | 3351 | |||
GenBank mRNA | FQ217109.1 | - | 1 | 1765 |
