siRNA Details
Gene Symbol: RGD1561444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242702.2 | XP_006242764.1 | 3 | 3147 | ||
XM_235936.8 | XP_235936.7 | 3 | 3147 | |||
GenBank mRNA | FQ217109.1 | - | 1 | 1561 |
