siRNA Details
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048215.1 | NP_001041680.1 | 14, 15 | 2062 | ||
XM_006242782.2 | XP_006242844.1 | 15, 16 | 2192 | |||
GenBank mRNA | DQ902828.1 | - | 14, 15 | 2062 |
