siRNA Details
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207610.1 | NP_997493.1 | 9 | 1360 | ||
XM_006242938.2 | XP_006243000.1 | 10 | 1574 | |||
GenBank mRNA | AY325198.1 | - | 9 | 1312 | ||
BC061761.1 | - | 9 | 1360 | 5621078 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145400, AK028721, AK028850, BC006649, AK172893.
Gene Ontology Categories:
