siRNA Details
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108144.1 | NP_001101614.1 | 3 | 211 | ||
XM_006243006.2 | XP_006243068.1 | 3 | 308 | |||
XM_006243007.2 | XP_006243069.1 | 3 | 308 | |||
XM_006243008.2 | XP_006243070.1 | 3 | 308 | |||
XM_006243009.2 | XP_006243071.1 | 3 | 308 | |||
XM_006243010.2 | XP_006243072.1 | 3 | 308 | |||
XM_006243011.2 | XP_006243073.1 | 3 | 308 | |||
XM_006243012.2 | XP_006243074.1 | 3 | 308 |
