siRNA Details
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127712.1 | NP_001121184.1 | 9, 10 | 1295 | ||
GenBank mRNA | AY325176.1 | - | 1067 | |||
BC133725.1 | - | 9, 10 | 1295 | 7100438 | ||
DQ083174.1 | - | 7, 8 | 959 |
