siRNA Details
Gene Symbol: Exph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236272.6 | XP_236272.4 | 6 | 5911 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Exph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236272.6 | XP_236272.4 | 6 | 5911 |