siRNA Details
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108174.1 | NP_001101644.1 | 4, 5 | 546 | ||
GenBank mRNA | BC160909.1 | - | 4, 5 | 504 | 7455732 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108174.1 | NP_001101644.1 | 4, 5 | 546 | ||
GenBank mRNA | BC160909.1 | - | 4, 5 | 504 | 7455732 |