siRNA Details
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729954.4 | XP_002730000.1 | 5 | 474 | ||
XM_006243554.1 | XP_006243616.1 | 6 | 622 | |||
XM_236501.7 | XP_236501.3 | 6 | 660 | |||
GenBank mRNA | FQ226547.1 | - | 6 | 627 |
