siRNA Details
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234266.2 | XP_006234328.1 | 10 | 1476 | ||
XM_236536.8 | XP_236536.6 | 10 | 1476 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234266.2 | XP_006234328.1 | 10 | 1476 | ||
XM_236536.8 | XP_236536.6 | 10 | 1476 |