siRNA Details
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234266.2 | XP_006234328.1 | 3 | 493 | ||
XM_236536.8 | XP_236536.6 | 3 | 493 | |||
GenBank mRNA | BC158847.1 | - | 3 | 440 | 7443919 |
