siRNA Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037784.1 | NP_001032873.1 | 2 | 294 | ||
XM_006234243.2 | XP_006234305.1 | 2 | 2509 | |||
XM_006234244.2 | XP_006234306.1 | 2 | 2508 | |||
GenBank mRNA | BC098943.1 | - | 2 | 294 | 7456980 |
