siRNA Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037784.1 | NP_001032873.1 | 3 | 480 | ||
XM_006234243.2 | XP_006234305.1 | 4 | 2803 | |||
XM_006234244.2 | XP_006234306.1 | 3 | 2694 | |||
XM_006234245.2 | XP_006234307.1 | 2 | 3131 | |||
XM_006234246.2 | XP_006234308.1 | 2 | 1382 | |||
GenBank mRNA | BC098943.1 | - | 3 | 480 | 7456980 |
