siRNA Details
Gene Symbol: Pls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108178.2 | NP_001101648.2 | 11 | 1240 | ||
XM_006243574.2 | XP_006243636.1 | 15 | 1843 | |||
XM_006243575.2 | XP_006243637.1 | 15 | 2223 | |||
XM_006243576.2 | XP_006243638.1 | 15 | 1913 | |||
XM_008766518.1 | XP_008764740.1 | 14 | 1857 | |||
XM_008766519.1 | XP_008764741.1 | 15 | 1763 | |||
GenBank mRNA | BC168847.1 | - | 11 | 1240 | 7446132 |
