siRNA Details
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108180.1 | NP_001101650.1 | 10 | 1351 | ||
XM_006243657.2 | XP_006243719.1 | 10 | 1393 | |||
XM_006243658.1 | XP_006243720.1 | 7 | 844 |
