siRNA Details
Gene Symbol: RGD1310507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047891.1 | NP_001041356.1 | 9, 10 | 1222 | ||
GenBank mRNA | AY325160.1 | - | 9, 10 | 1139 | ||
AY539912.1 | - | 9, 10 | 1222 |
