siRNA Details
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244011.2 | XP_006244073.1 | 26 | 3917 | ||
XM_006244013.2 | XP_006244075.1 | 27 | 3749 | |||
XM_006244014.2 | XP_006244076.1 | 26 | 3948 | |||
XM_006244015.2 | XP_006244077.1 | 27 | 4004 | |||
XM_006244016.2 | XP_006244078.1 | 27 | 4001 | |||
XM_006244018.2 | XP_006244080.1 | 26 | 4124 | |||
XM_008766654.1 | XP_008764876.1 | 26 | 3951 | |||
XM_008766655.1 | XP_008764877.1 | 16 | 1842 | |||
XM_236654.8 | XP_236654.6 | 26 | 4158 |
