siRNA Details
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244011.2 | XP_006244073.1 | 4 | 1474 | ||
XM_006244013.2 | XP_006244075.1 | 5 | 1306 | |||
XM_006244014.2 | XP_006244076.1 | 4 | 1505 | |||
XM_006244015.2 | XP_006244077.1 | 5 | 1561 | |||
XM_006244016.2 | XP_006244078.1 | 5 | 1558 | |||
XM_006244018.2 | XP_006244080.1 | 4 | 1681 | |||
XM_008766654.1 | XP_008764876.1 | 4 | 1508 | |||
XM_236654.8 | XP_236654.6 | 4 | 1715 |
