siRNA Details
Gene Symbol: Pomgnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009437.1 | NP_001009437.1 | 1 | 1085 | ||
XM_008766681.1 | XP_008764903.1 | 2 | 1357 | |||
XM_008766682.1 | XP_008764904.1 | 2 | 2722 | |||
XM_008766683.1 | XP_008764905.1 | 3 | 1427 | |||
GenBank mRNA | AJ868534.1 | - | 1 | 1085 |
