siRNA Details
Gene Symbol: Exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100725.1 | NP_001094195.1 | 2 | 135 | ||
GenBank mRNA | BC089995.1 | - | 1 | 26 | ||
BF561514.1 | - | 2 | 141 | |||
FQ226090.1 | - | 2 | 140 |
