siRNA Details
Gene Symbol: Pp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134567.1 | NP_001128039.1 | 2 | 811 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134567.1 | NP_001128039.1 | 2 | 811 |