siRNA Details
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108199.1 | NP_001101669.1 | 9 | 2760 | ||
XM_006244452.2 | XP_006244514.1 | 14 | 3614 | |||
XM_006244453.2 | XP_006244515.1 | 14 | 3554 | |||
XM_008766863.1 | XP_008765085.1 | 13 | 3527 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172622, NM_001097623, AK046822, BC051645, BC108416, and others.
Gene Ontology Categories:
