siRNA Details
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191582.1 | NP_001178511.1 | 12, 13 | 3039 | ||
XM_006244518.2 | XP_006244580.1 | 12, 13 | 3363 | |||
XM_008766864.1 | XP_008765086.1 | 12, 13 | 3363 |
