siRNA Details
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108203.1 | NP_001101673.1 | 12 | 1198 | ||
XM_006244524.2 | XP_006244586.1 | 12 | 1194 | |||
XM_006244525.2 | XP_006244587.1 | 12 | 1191 | |||
XM_006244526.2 | XP_006244588.1 | 11 | 1064 | |||
XM_006244527.2 | XP_006244589.1 | 11 | 1096 | |||
XM_006244528.2 | XP_006244590.1 | 11 | 1093 | |||
XM_006244529.2 | XP_006244591.1 | 10 | 966 | |||
XM_006244530.2 | XP_006244592.1 | 10 | 963 | |||
XM_006244532.2 | XP_006244594.1 | 11 | 1091 | |||
XM_006244533.2 | XP_006244595.1 | 10 | 961 | |||
XM_006244534.2 | XP_006244596.1 | 11 | 1164 | |||
XM_006244535.2 | XP_006244597.1 | 9 | 1459 | |||
GenBank mRNA | FQ221753.1 | - | 9 | 1440 | ||
FQ221813.1 | - | 9 | 1829 |
