siRNA Details
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108214.2 | NP_001101684.2 | 3 | 623 | ||
XM_008767031.1 | XP_008765253.1 | 2 | 98 | |||
XM_008767032.1 | XP_008765254.1 | 3 | 1349 | |||
XM_008767033.1 | XP_008765255.1 | 3 | 813 | |||
XM_008767034.1 | XP_008765256.1 | 3 | 1706 |
