siRNA Details
Gene Symbol: Tmem237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108220.1 | NP_001101690.1 | 11 | 1173 | ||
XM_006244975.2 | XP_006245037.1 | 12 | 1872 | |||
XM_006244976.2 | XP_006245038.1 | 12 | 1497 | |||
XM_006244977.2 | XP_006245039.1 | 12 | 2113 | |||
XM_008767098.1 | XP_008765320.1 | 12 | 1885 |
